Would you like to log in again?
PurSwab® ESD Foam Tipped Swabs, Static-Dissipative Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
1621-PFESD 1806-PFESD 1621-PFESD 1273-PFESD 1806-PFESD 1273-PFESD 1283-PF-ESD 1283-PF-ESD
89108-136CS 1189.67 USD InStock
89108-136 89087-860 CA89108-136 CA89087-862 89087-862 CA89087-860 CA89087-858 89087-858
PurSwab® ESD Foam Tipped Swabs, Static-Dissipative Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Clinical Use
These swabs are designed for working in and around the most static-sensitive components and parts.
A range of shapes, sizes, and materials allow users to find a swab made specifically for any application. Made in USA.
Certifications: CE marked.
Ordering information: Swabs are packaged 50 swabs per bag, 20 bags per case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty