Would you like to log in again?
Process NPD® Disinfectant, STERIS®
Supplier: Steris
Process NPD®
639008 160528
89087-880CS 754.9 USD InStock
89087-880 21899-728
Process NPD® Disinfectant, STERIS®
Cleaning Agents Disinfectants
This phosphate-free, quaternary ammonium germicide is a one-step, detergent-based disinfectant formulated to provide broad-spectrum coverage on hard, nonporous surfaces
  • Dilute 1:256
  • Filtered double bagged, gamma irradiated, and lot traceable
Cleaner contains no perfumes or dyes that could interfere with critical research studies or manufacturing processes.
Unit dose pouch produces 2 gallon of solution after dilution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty