Would you like to log in again?
Memphis UltraTech® Gloves, MCR Safety
Supplier: MCR Safety
Memphis UltraTech®
9693L 9693XL 9693M 9693XS 9693S
89087-952PK 281.93 USD InStock
89087-952 80081-722 80081-720 80081-726 80081-724
Memphis UltraTech® Gloves, MCR Safety
Gloves
Cut protection can be felt.
  • Treated with Actifresh® to help prevent order and bacteria
  • Cut protection plus dexterity is what needed to complete the task
This 13 gauge stretch Kevlar® shell and light foam nitrile palm and fingertips gives a softer and more flexible option in cut protection.
Ordering information: These gloves are polybagged by the pair.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty