Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hypochlorite 4 - 6% in aqueous solution

Supplier: HAWKINS CHEMICAL CO
Danger

32125-DFS 32124-DFS 32184-DFS
89087-980EA 425 USD InStock
89087-980 89087-982 89087-978
Sodium hypochlorite 4 - 6% in aqueous solution
Sodium hypochlorite
Extremely low mercury.


Formula: ClNaO
MW: 74.44 g/mol
Boiling Pt: 111 °C (1013 hPa)
Melting Pt: –30…–20 °C
Density: 1.22…1.25 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty