Would you like to log in again?
Polynit Z Wipes, Contec
Supplier: Contec
Polynit-Z
LWZS0001
89089-312CS 669.23 USD InStock
89089-312
Polynit Z Wipes, Contec
Wipes Cleanroom Wipes Dry Cleanroom Wipes
The Polynit Z Wipes are constructed using conductive yarns within the polyester interlock knit that feature a unidirectional conductive pattern. These are very clean static dissipative wipes that is well suited for applications sensitive to electrostatic discharge.
  • Recommended for ISO Class 3-5 environments
  • Polyester fabric is extremely low in both particles and extractables
  • Good sorbency makes it compatible with many solvents
  • Polyester fabric is abrasion and chemical resistant
These are laundered and packed in an ISO Class 4 cleanroom environment. The laser-cut edges seal fibers at the wipes edge. Ideal for cleaning the interior surface of process tools and other equipment.
Average surface resistivity: 3.5×10¹⁰ Ω/sq.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty