Would you like to log in again?
Sterile SatPax® MicroSeal® WFI Presaturated Nonwoven Cleanroom Wiper, Berkshire
Supplier: Berkshire
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SatPax®
SSPMS01.5WFI
89089-432CS 1048.19 USD InStock
89089-432
Sterile SatPax® MicroSeal® WFI Presaturated Nonwoven Cleanroom Wiper, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Sterile SatPax® MicroSeal® WFI combines MicroSeal® 1200 ultrasonically sealed-edge cleanroom laundered wipers composed of 100% continuous filament polyester knit fabric with a high saturation level of 70% IPA and 30% WFI. This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents.
  • 100% continuous filament polyester knit
  • Ultrasonically sealed edge for reduced fiber contamination
  • Laundered and packaged in Berkshire’s ISO Class 4 cleanroom
  • Triple packaged for easy entry into sterile environments
  • Re-sealable solvent resistant packaging
Solvent is 70% isopropyl alcohol and 30% water for injection (WFI). No chemical binders in base material. Suitable for ISO Class 3 (FED-STD-209E Class 1/M1.5) and less critical environments. Excellent for wet cleaning of critical surfaces where control of flammable solvents and flammable solvent concentrations is required.
Certifications: Gamma irradiated to a Sterility Assurance Level of 10⁻⁶ per AAMI guidelines. Certificate of sterility. Passes LAL test for endotoxins <20 EU/device. Complies with RoHS and REACH requirements. Meets cGMP requirements for traceability. Laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Ordering information: Packaged in resealable, solvent-resistant packaging.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty