Would you like to log in again?
SP Bel-Art Bel-Art Bulk-Packed Elliptical (Egg-Shaped) Spinbar Magnetic Stirring Bars, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
37130-9001 37130-9038 37130-9004 37130-9058 37130-9007 37130-9008 37130-9006
10767-156CS 1070.37 USD InStock
10767-156 89089-568 89089-564 89089-574 10767-160 89089-572 89089-570
SP Bel-Art Bel-Art Bulk-Packed Elliptical (Egg-Shaped) Spinbar Magnetic Stirring Bars, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
These egg-shaped stirring bars are designed especially for use in round-bottom vessels as they spin unrestricted by the vessel contour.
  • Particularly well suited for round bottom flasks; shape ensures complete mixing
  • Offers minimal contact when used in plastic containers
  • Egg shape provides pivot point without a separate ring or abrupt change in contour
  • Autoclave safe
Magnet itself is egg-shaped encapsulated in FDA grade, USP Class VI Teflon® PTFE, except Models 37130-9038, 37130-9004, and 37130-9058 which have a cylindrical magnet in egg-shaped Teflon® PTFE.
Certifications: Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty