Would you like to log in again?
Corning® Screw Cap with Plug Seal, Polypropylene, Corning
Supplier: Corning
Corning®
1397-80LTC
89089-662CS 126.6 USD InStock
89089-662
Corning® Screw Cap with Plug Seal, Polypropylene, Corning
Closures Bottle Closures
Linerless orange cap has GLS80 thread and is autoclavable at 121°C (250°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty