Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Corning® 120 mm Center Cap Assembly for 3 L to 36 L Proculture® Spinner Flasks, Corning

Supplier: Corning

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Corning®
402577 402576 402579
89089-674CS 913.88 USD InStock
89089-674 89089-672 89089-670
Corning® 120 mm Center Cap Assembly for 3 L to 36 L Proculture® Spinner Flasks, Corning
Closures Bottle Closures
120 mm center cap assembly for Proculture® Spinner Flasks.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty