Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Corning® Disposable Aseptic Transfer Cap for Disposable Spinner Flask, Corning

Supplier: Corning

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Corning®
3558 3562 3565 3545 3564 3528 3567
89089-914CS 47.82 USD InStock
89089-914 89176-434 89089-912 89089-910 89089-908 89176-436 89176-438
Corning® Disposable Aseptic Transfer Cap for Disposable Spinner Flask, Corning
Closures Bottle Closures
These disposable Corning® spinner flask aseptic transfer caps will both reduce contamination and enhance the productivity of your cell culture operations when adding or removing medium and cells.

  • Cap is made of high density polyethylene; autoclaving is not recommended
  • The venting port ends in a 25 mm diameter vent filter with a 0.2 µm pore PVDF membrane and a barbed fitting
  • Chemically resistant, heat sealable flexible tubing, ending in a male medical plastic coupler (MPC) connector closed with a female sealing cap

Aseptic transfer caps are sterile (10⁻⁶ SAL), individually double bagged and fit the side arms on 1 L Corning® disposable spinner flasks (Cat. No. 89089-754). Cap is made of high density polyethylene; autoclaving is not recommended. The venting port ends in a 25 mm diameter vent filter with a 0.2 µm pore PVDF membrane and a barbed fitting. The liquid transfer port has an internal end with a rigid polypropylene dip tube angled to reach the bottom of the flask and on the external side a ¹/₈" ID (3.2 mm), ¹/₄" OD chemically resistant, heat sealable flexible tubing, ending in a male medical plastic coupler (MPC) connector closed with a female sealing cap.

Packaging: Aseptic transfer caps are sterile (10⁻⁶ SAL), individually double bagged.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty