Would you like to log in again?
Corning® Pipette Tips, Universal, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
4860 4806 4868 4862 4865 4864 4710 4844 4712 9032 4863 4713 4846 4866 4867 4804 4715 4714 4845 4711 4803
29442-658CS 569.8 USD InStock
29442-658 29442-688 29442-656 29442-686 29442-654 29442-684 29443-350 29443-352 89089-708 29442-706 29443-270 89089-706 89089-958 29443-348 89089-956 89089-714 89089-712 29442-682 29442-680 29442-692 29442-690
Corning® Pipette Tips, Universal, Corning
Pipette Tips
Corning® universal fit tips are designed to provide a reliable fit with all major brand pipettors.
  • Beveled orifice helps ensure accurate fluid delivery
  • All tips are certified RNase-/DNase-free and non pyrogenic
  • Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
  • Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty