Would you like to log in again?
PYREX® VISTA™ Bulb Pipettes, Class A, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PYREX® VISTA™
70710-1 70710-10 70710-5 70710-2
89090-170CS 167.09 USD InStock
89090-170 89090-168 89090-164 89090-166
PYREX® VISTA™ Bulb Pipettes, Class A, Corning
Pipettes Graduated Pipettes
Reusable.
These bulb pipettes are manufactured to Class A capacity tolerances as indicated by ASTM E-969. The pipettes are color-coded according to their capacity and feature colored markings.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty