Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Corning® Magnetic Stir Bar Retriever, Corning
Supplier: Corning
Corning®
6970SR
89090-234CS 41.87 USD InStock
89090-234
Corning® Magnetic Stir Bar Retriever, Corning
Stirrers Stirring Tools
The stir bar retriever contains a strong magnet to remove a stir bar from all glass and plastic containers used in the laboratory.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty