Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Petri dishes, soda lime silca glass, Corning®
Supplier: Corning
Corning®
70165-60 70165-100 70165-152 70165-101 70165-102
89090-270CS 138.22 USD InStock
89090-270 89090-272 89090-278 89090-274 89090-276
Petri dishes, soda lime silca glass, Corning®
Culture Dishes Petri Dishes
Flat, clear petri dishes are manufactured from soda lime silica glass and will withstand repeated sterilization (wet or dry)
Edges are beaded to provide greater mechanical strength. The bead also provides a means to equally space the side walls of the bottom and cover, reducing capillary action of any condensed moisture within. Dish bottoms feature a triangular enamel reference point for serial dilutions. Cover does not provide airtight seal. Chemical and thermal-resistant.
Caution: These dishes are not designed for rapid thermal shock.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty