Would you like to log in again?
PYREXPLUS® Plastic-Coated Filtering Flasks, Graduated, Heavy Wall, Corning
Supplier: Corning
PYREXPLUS®
65340-500 65340-250 65340-1L 65340-4L 65340-2L
29410-988CS 338.63 USD InStock
29410-988 89091-694 89090-588 89091-676 89090-584
PYREXPLUS® Plastic-Coated Filtering Flasks, Graduated, Heavy Wall, Corning
Flasks Filter Flasks
PYREXPLUS® coated.
  • Resistant to thermal shock
  • Graduated to show approximate volume
  • To allow easy connection of standard size tubing, side arm is not coated
These coated heavy wall flasks have protective PVC coating for longer product life and safety. The coating makes cleanup faster and easier by reducing spill volume if breakage occurs. Coating is autoclavable to 121 °C (250 °F). Do not heat above 121 °C (250 °F) using moist heat or 110 °C (230 °F) using dry heat.
Caution: Caution: Do not place plastic-coated ware over direct heat or flames.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty