Would you like to log in again?
PYREXPLUS® Graduated Cylinders, To Deliver, Corning
Supplier: Corning
PYREXPLUS®
63024-500 63024-100 63024-2L 63024-1L 63024-250
89090-590CS 506.88 USD InStock
89090-590 89090-580 89090-586 89090-578 89090-582
PYREXPLUS® Graduated Cylinders, To Deliver, Corning
Cylinders Measuring Cylinders
Borosilicate glass.
  • Calibrated to deliver (TD)
  • Graduations and markings in durable white enamel
  • Resistant to thermal shock
Cylinder features a protective polymer coating for longer product life and safety.
Coating makes cleanup faster and easier by reducing spill volume if breakage does occur. Coating is autoclavable to 121°C (249.8°F). Do not heat above 121°C (249.8°F) moist heat or 110°C (230°F) dry heat. Do not place coated ware over direct heat or open flames. Can be written on with any standard lab marker, permanent or water-soluble. Cylinder features single metric scale. Bumper guards not included.
PYREXPLUS® labware allows for a better hand grip and clamp hold.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty