Would you like to log in again?
PYREX® 105° Angle Vacuum Connecting Adapter with 24/40 Joints, Corning
Supplier: Corning
PYREX®
8946-24
89090-630CS 1078.32 USD InStock
89090-630
PYREX® 105° Angle Vacuum Connecting Adapter with 24/40 Joints, Corning
Distillation Components Distillation Adapters
This distillation adapter tube with sidearm has an outer joint at the top and an inner-sealed through drip tip joint at the bottom.
  • 105° angle
  • Top [ST] outer joint
  • Bottom drip tip [ST] joint
  • Serrated sidearm
Adapter tube features a [ST] 24/40 outer joint at the top and an inner-sealed through drip tip [ST] 24/40 joint at the bottom. The serrated sidearm is 10 mm OD. Drip tip at the bottom extends 20 mm below the lower end of the joint.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty