Would you like to log in again?
PYREX® Volumetric Flasks, Class A, PTFE Stopper, Corning
Supplier: Corning
PYREX®
5644-100 5644-25 5644-200 5644-50 5644-250 5644-500 5644-2L 5644-10 5644-1L
89090-816CS 1148.13 USD InStock
89090-816 89090-678 89090-910 89090-712 89090-768 89090-845 89090-604 89091-100 89090-750
PYREX® Volumetric Flasks, Class A, PTFE Stopper, Corning
Flasks Volumetric Flasks
These class A PYREX® volumetric flasks have machine-blown bodies to which are sealed heavy-beaded, heavy-tubing necks, tooled for PTFE color-coded keyhole stoppers
  • Strength increased through machine-blown bodies
  • Sharp graduation line and permanent and large white block letters make the inscription easy to read
The graduation line is sharp and permanent. Large white block letters make the inscription easy to read. These flasks have been manufactured to Class A tolerances as established by ASTM E-694 for volumetric ware, ASTM E-542 for calibration of volumetric ware, and ASTM E-288 for volumetric flasks.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty