Would you like to log in again?
PYREX® Burets, Class B, Fluoropolymer Resin Stopcock, Corning
Supplier: Corning
PYREX®
2116-25 2116-50
89090-914CS 977.64 USD InStock
89090-914 89090-820
PYREX® Burets, Class B, Fluoropolymer Resin Stopcock, Corning
Burets
Made of PYREX® borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Replaceable stopcock
  • White enamel graduations
These burets are designed to reduce replacement costs by using replaceable components. These sturdy burets have tooled tops and bottoms. The replaceable screw thread locks the PTFE stopcock with ground tip.
Ordering information: Buret shipped complete and ready-to-use with dust cover and 2 mm fluoropolymer resin stopcock.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty