Would you like to log in again?
Corning® Frosted Micro Slides, Corning
Supplier: Corning
Corning®
2948-75X25 2949-75X25
89091-004CS 621.87 USD InStock
89091-004 48312-048
Corning® Frosted Micro Slides, Corning
Slides Microscope Slides
For use in nonsterile cytology and pathology applications.
Made of water-white glass for maximum clarity. Precleaned and ready for use. Inspected for chips and rough edges before packaging. Slides are sandblasted and not etched, resulting in high legibility against most laboratory bench backgrounds.
Ordering information: Packaged 1/2 gross per box. Slides are packed vertically for easy lift out.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty