Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PYREX® Oil Conical Centrifuge Tube 100 ml Lifetime-Red™ with White Graduations, Corning

Supplier: Corning

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PYREX®
8180-100
89091-072CS 1210.56 USD InStock
89091-072
PYREX® Oil Conical Centrifuge Tube 100 ml Lifetime-Red™ with White Graduations, Corning
Tubes Centrifuge Tubes
These 100 ml PYREX® conical centrifuge tubes are used in accordance with standard methods of testing for water and sediment in petroleum products.

They have white graduations against a permanent red panel for easier reading. Graduated upward to 0.5 ml in divisions of 0.05 ml; from 0.5 to 2 ml in divisions of 0.1 ml; from 2 to 3 ml in divisions of 0.2 ml; from 3 to 5 ml in divisions of 0.5 ml; from 5 to 10 ml in divisions of 1 ml; from 10 to 25 ml in divisions of 5 ml and at 50, 75 and 100 ml.

Certifications: ASTM D-96, D-91, D-893 and D-1796.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty