Would you like to log in again?
PYREX® Volumetric Pipettes, Class A, Color-Coded, Graduated, Serialized/Certified, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PYREX®
7101-25 7101-50 7101-15 7101-100 7101-1 7101-2 7101-3 7101-5 7101-20 7101-4 7101-10
89091-226CS 176.71 USD InStock
89091-226 89091-304 89091-392 89091-082 89091-270 89091-490 89091-364 89091-122 89091-466 89091-178 89091-234
PYREX® Volumetric Pipettes, Class A, Color-Coded, Graduated, Serialized/Certified, Corning
Pipettes Graduated Pipettes
Pipettes are calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969.
  • Sizes 1ml through 25ml are color-coded for ease in sorting and selection
  • Colored graduations are enameled on to the glass
PYREX® Serialized/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Delivery information: Each pipette is individually serialized and supplied with a certificate of identity and capacity, traceable to NIST standards.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty