Would you like to log in again?
PYREX® 105° Vacuum Connecting Adapters with [ST] Joints, Corning
Supplier: Corning
PYREX®
8947-14 8947-19
32645-323CS 176.51 USD InStock
32645-323 89091-086
PYREX® 105° Vacuum Connecting Adapters with [ST] Joints, Corning
Adapters Adapters, Joints with Tube Connectors
This distillation adapter tube with sidearm has an outer joint at the top and an inner-sealed through drip tip joint at the bottom.
  • 105° angle
  • Top [ST] outer joint
  • Bottom drip tip [ST] joint
  • Serrated sidearm
Adapter tube features a [ST] outer joint at the top and an inner-sealed through drip tip [ST] joint at the bottom. The serrated sidearm is 10 mm OD.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty