Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PYREX® 105° Angled Tube Adapter with 24/40 Standard Taper Joint , Corning

Supplier: Corning
PYREX®
8840-24
89091-088CS 690.5 USD InStock
89091-088
PYREX® 105° Angled Tube Adapter with 24/40 Standard Taper Joint , Corning
Adapters Adapters, Joints with Tube Connectors
These angled tube adapters are made of PYREX® borosilicate glass.

  • Arms are approximately 105° apart
  • Upper end equipped with an outer 24/40 standard taper joint
  • Lower end is cut off at an angle and is used as a receiver adapter

Tube O.D. is 12 mm.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty