Would you like to log in again?
PYREX® Graduates, Beakers Form, Double Spout, Double Scale
Supplier: Corning
PYREX®
6480-125 6480-1L 6480-250 6480-500 6480-2L
89090-716CS 955.87 USD InStock
89090-716 89091-406 89090-850 89091-130 89091-246
PYREX® Graduates, Beakers Form, Double Spout, Double Scale
Beakers
PYREX®, borosilicate glass, double spout, double scale beakers.
  • Increased wall thickness for better mechanical properties
  • Ideal for measuring hot solutions
  • Should not have direct contact with heat source
Thick walled, with spout, approximate graduations for both fluid ounces and milliliters.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty