Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PYREX® Boiling Flasks, Flat Bottom, Short Neck, Standard Taper Joint, Corning
Supplier: Corning
PYREX®
4100-250 4100-125 4100-1L 4100-500 4100-50
89090-796CS 557.62 USD InStock
89090-796 29113-060 89091-136 89090-692 89090-882
PYREX® Boiling Flasks, Flat Bottom, Short Neck, Standard Taper Joint, Corning
Flasks Flat Bottom Flasks
PYREX® borosilicate glass, clear, with short neck.
  • High thermal shock resistance
  • Chemically inert
  • Provide contamination free results
  • Long life
These flasks have full length outer standard tapered 24/40 joint. The normal working temperature is 230 °C (446 °F).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty