Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PYREX® Heavy Duty Conical Centrifuge Tubes, Graduated, Corning

Supplier: Corning

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PYREX®
8140-40 8140-12
89091-378CS 595.57 USD InStock
89091-378 89091-146
PYREX® Heavy Duty Conical Centrifuge Tubes, Graduated, Corning
Tubes Centrifuge Tubes
PYREX® borosilicate glass, conical, heavy duty, beaded rim.

  • Tubes 12 ml are graduated upward from tip in 0.1 ml divisions
  • Tubes 40 ml are graduated in 0.5 ml divisions to 10 ml and from 10 to 40 ml in 1 ml divisions

The tubes are of heavy wall construction, for greater mechanical strength, with durable, white enamel graduations.

In order to conform to outside diameter and overall length limitations imposed by centrifuge rotors and supports, it has been necessary to reduce the capacity of these heavy duty tubes.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty