Would you like to log in again?
PYREX® Conical Centrifuge Tubes with Black Graduations, Beaded Rim, Corning
Supplier: Corning
PYREX®
8101-15 8101-50
89091-170CS 351.96 USD InStock
89091-170 21053-140
PYREX® Conical Centrifuge Tubes with Black Graduations, Beaded Rim, Corning
Tubes Centrifuge Tubes
These tubes have sharp, colored enamel graduations and markings.
The tubes are the same size and shape as Cat. Nos. 8060 and 8080. They have precision formed tips and uniform walls throughout for a high degree of accuracy and mechanical strength.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty