Would you like to log in again?
PYREX® Filtering Flasks, Heavy Wall, Replaceable Tubulation, Corning
Supplier: Corning
PYREX®
5341-1L 5341-250 5341-2L 5341-500 5341-4L
89091-290CS 1316.34 USD InStock
89091-290 89090-734 89091-200 29416-306 29416-328
PYREX® Filtering Flasks, Heavy Wall, Replaceable Tubulation, Corning
Flasks Filter Flasks
PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These filtering flask have bent, poly tubulation in a grommet which acts as a shock absorber. Graduated to show approximate capacity.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty