Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
PYREX® Distilling Flasks, Angle Type, 3 Necks, [ST] Joints, Corning
Supplier: Corning
PYREX®
4965B-250 4965A-500 4965A-1L 4965-500 4965-1L 4965-100 4965B-500
89091-080CS 812.96 USD InStock
89091-080 89091-216 29281-131 89091-430 32645-390 32645-392 32645-391
PYREX® Distilling Flasks, Angle Type, 3 Necks, [ST] Joints, Corning
Flasks Distillation Flasks
PYREX® borosilicate glass, with angled side necks, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One center neck and two side necks. Three necks with two angled side necks at the same height with 24/40 standard taper outer joints and a larger center neck with a vertical 34/45 standard taper joint. Matt white labelling area.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty