Would you like to log in again?
PYREX® Conical Centrifuge Tubes with White Graduations and Standard Taper Pennyhead Stopper, Corning
Supplier: Corning
PYREX®
8084-5 8084-50 8084-15
89091-172CS 712.7 USD InStock
89091-172 89091-440 89091-482
PYREX® Conical Centrifuge Tubes with White Graduations and Standard Taper Pennyhead Stopper, Corning
Tubes Centrifuge Tubes
PYREX®, borosilicate glass.
These tubes have white enamel graduations. The 5 and 15 ml sizes are graduated upward from the tip in 0.1 ml increments. The 50 ml size is graduated upward from tip to 10 ml in 0.5 ml increments, and from 10 ml to 50 in 1 ml increments. The pennyhead stopper is solid.
For stopper only, see Cat. No. 7661.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty