Would you like to log in again?
PYREX® Micro Burets, Class A, Funnel Top, Precision Bore, Straight [ST] Stopcock, Corning
Supplier: Corning
PYREX®
2128-5 2128-10
89091-092CS 1298.61 USD InStock
89091-092 89091-478
PYREX® Micro Burets, Class A, Funnel Top, Precision Bore, Straight [ST] Stopcock, Corning
Burets
Micro burets, Class A, PYREX® borosilicate glass.
  • Precision bore tubing
  • Funnel top
  • Tapered tips for smooth flow
  • Stopcocks turn freely yet give positive control
Designed for precise analytical methods in microchemistry. 
Certifications: The capacity tolerance on these burets is established by ASTM E1189 and are calibrated in accordance with ASTM E542.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty