Would you like to log in again?
Air Flow Indicator Kit, Sensidyne
Supplier: Sensidyne
7014094
89091-766EA 282.15 USD InStock
89091-766
Air Flow Indicator Kit, Sensidyne
Anemometers
This Air Flow Indicator (HVAC) Kit is highly recommended to track air flow currents and for fit testing respirators per OSHA 29 CFR part 1910.134, irritant fume protocol.
Delivery information: The kit includes carrying pouch, aspirator bulb, tube tip breaker, and a box of smoke tubes.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty