Would you like to log in again?
Nalgene® Teflon® Fluorinated Ethylene Propylene, Low Particulate and Low Metals Bottles, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
381600-0008 381600-0016 381600-0032 381600-0032 381600-0004
89091-778CS 979.2 USD InStock
89091-778 89091-782 89091-780 CA89091-784 89091-784
Nalgene® Teflon® Fluorinated Ethylene Propylene, Low Particulate and Low Metals Bottles, Thermo Scientific
Bottles Plastic Bottles
Nalgene® Teflon® fluorinated ethylene propylene low particulate and low metals bottles are narrow-mouth bottles with particle levels of less than 20 particles per ml at 0.3 µm and greater.
  • Very good chemical resistance
  • Good temperature resistance
  • Linerless closure
Each bottle is double bagged under Class 10 laminar flow hoods inside a Class 100 clean room. The bottles are excellent for storing high-purity chemicals and include a certificate of quality that assures the products have been tested and accepted in accordance with specifications.
Certifications: Metals certified to (µg/L) ppb levels <0.20 Hg, <0.5 Be, <1.0 As, Cd, Pb, <2.0 Sb, Se, <5.0 Ag, Co, Cr, Cu, Mn, Th, V, <10 Ba, Ni, Zn, <50 Mg, <75 Al, <100 Ca, Fe, K, Na.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty