Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunc® IVF Four-Well Dishes, Thermo Scientific
Supplier: Thermo Scientific
Nunc®
144444 144444 179830 179830
73521-354CS 609.66 USD InStock
73521-354 CA73521-354 89092-330 CA89092-330
Nunc® IVF Four-Well Dishes, Thermo Scientific
Culture Dishes Petri Dishes
Four-well multidishes are for use in human In Vitro Fertilization (IVF) and provide security, safety, and reproducibility for valuable samples.
Dishes ensure proper orientation and prevent cross-contamination with the notched lid and dish. Prevent scratches on the well bottoms and maintain an optimal temperature with a 0.8mm raised edge.
Certifications: Produced under ISO 13485. MEA tested. CE marked class II medical device and FDA cleared for use in IVF. Uniquely certified based on a one-cell stage mouse embryo toxicity test. Stringent lot-level manufacturing controls ensure viability, reliability, and reproducibility.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty