Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunc® IVF Petri Dishes, Thermo Scientific
Supplier: Thermo Scientific
Nunc®
150360 150255 150270 150255 150360
89092-336CS 427.1 USD InStock
89092-336 CA89092-336 89092-332 89092-334 CA89092-332
Nunc® IVF Petri Dishes, Thermo Scientific
Culture Dishes Petri Dishes
Thermo Scientific Nunc® provide an extensive portfolio of products intended for use in human In Vitro Fertilization (IVF)
The Nunc® IVF Petri Dish facilitates gas exchange with a vented lid, and comes in a variety of sizes for your applications.
Thermo Scientific Nunc® InVitro Fertilization Products provide security, safety and reproducibility for your valuable samples, and are uniquely certified based on a 1-cell stage mouse embryo toxicity test. Stringent lot-level manufacturing controls ensure viability, reliability and reproducibility.
Products are produced under ISO 13485, MEA tested, CE marked class II medical device and FDA cleared for use in IVF.
Certifications: CE and FDA
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty