Would you like to log in again?
Roll-O-Matic® Self-Wringing Cleanroom Sponge Mops, Vileda Professional-FHP
Supplier: Vileda Professional
118549 118535 126104 122910 118550 120848
94002-800EA 433.13 USD InStock
94002-800 94001-380 68400-492 94002-802 89209-458 89092-938
Roll-O-Matic® Self-Wringing Cleanroom Sponge Mops, Vileda Professional-FHP
Mops
The ideal self-wringing cleanroom mop
  • Straight handles allow user to properly use the pull-and-lift technique with both sides of the mop
  • Perfect for use on floors, walls, and ceilings
  • Ergonomic grips reduce worker fatigue
  • Entire aluminum and steel mop handles are completely autoclavable at the most common autoclave cycles
  • Caps on steel versions are stainless steel and completely sealed
  • Aluminum version features color coding tabs (blue, red, green, yellow) for easy equipment identification
The Vileda Professional® stainless steel and aluminum extendable Roll-O-Matic® mops are the best choice when seeking one mop to apply disinfectant, clean surfaces, or absorb spills in all production and controlled environments. These stainless steel Roll-O-Matic® mops are manufactured to resist repeated autoclave cycles and continued exposure to the common chemicals and disinfectants used in the controlled environment marketplace.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty