Would you like to log in again?
KIMTECH PURE® G3 STERLING™ Nitrile Gloves, Ambidextrous, Kimberly-Clark
Supplier: Kimberly-Clark
Kimtech™ Sterling™
99235 99239 99238 99237 99238 99234 99236 99235
89093-002CS 1005.68 USD InStock
CA89092-994 89093-002 CA89093-000 89092-996 89093-000 CA89092-998 CA89092-992 89092-994
KIMTECH PURE® G3 STERLING™ Nitrile Gloves, Ambidextrous, Kimberly-Clark
Gloves
Powder-free, latex-free gloves provide the protection of nitrile and sensitivity comparable to natural rubber latex.
  • Excellent tactile sensitivity with superior tensile strength
  • Static-dissipative in use
Suitable for use in the manufacture of pharmaceuticals, medical devices, electronics, disk drives, flat panel displays, and semiconductors. Silicone-free. Ambidextrous. Beaded cuff. Textured fingertips. AQL 1.5. Packaged in a Class 100 cleanroom.
Certifications: Recommended for ISO Class 4 (FED-STD-209E Class 1/M1.5) or higher cleanroom environments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty