Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 25% (w/w) in aqueous solution, food grade

Supplier: Columbus Chemical Industries, Inc
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

964200-21901
89093-148EA 589.43 USD InStock
89093-148
Sodium hydroxide 25% (w/w) in aqueous solution, food grade
Sodium hydroxide
Synonyms: Caustic soda solution


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty