Would you like to log in again?
Nalgene® PETG Certified Clean Containers, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
382019-2000 382019-0030 382019-0250 382019-1000 382019-0060 382019-0500 382019-0125
89093-200CS 712.66 USD InStock
89093-200 89093-198 89093-196 89093-194 89093-206 89093-204 89093-202
Nalgene® PETG Certified Clean Containers, Thermo Scientific
Bottles Plastic Bottles
Nalgene® PETG Certified Clean Containers are suitable for pharmaceutical and biotechnology applications requiring the processing and storage of bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations.
The packaging is designed for cleanroom applications. The bottle and closure are secured with heat-shrink bands providing a tamper-evident seal and then packaged in cleanroom grade heat-sealed bags to assure product integrity. They are then gamma irradiated to provide a 10⁻⁶ sterility assurance level (SAL). Bottles meet the requirements for USP Class VI, EP Modified Abnormal Toxicity Test, are non-cytotoxic, non-pyrogenic, non-hemolytic, and comply with USP <661> physicochemical guidelines.
The materials of construction provide excellent gas barrier properties to minimize pH shifts. The resins used to produce these containers are free of animal derived components (ADCF). The bottle and closure system is guaranteed leakproof, making it suitable for transportation applications.
Certifications: Manufactured in a certified ISO 14644-1 Class 7 cleanroom, these containers are lot certified to comply with particulate limits specified in USP <788>, EP 2.9.19 and JP 14th edition Part 1 Section 24.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty