Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
BRAND BRANDplates® cellGrade™ plus Microplates for Cell Culture, Sterile, BrandTech®
Supplier: BrandTech
BRANDplates®
782030 782022 782031
89093-692PK 519.96 USD InStock
89093-692 89093-700 89093-698
BRAND BRANDplates® cellGrade™ plus Microplates for Cell Culture, Sterile, BrandTech®
Microplates Cell Culture Plates
BRANDplates® cellGrade™ plus plates are sterile plates treated for the cultivation of fastidious adherent cell lines
  • Enhanced treatment for fastidious cell lines
  • ANSI-SBS formatted
  • Sterility assurance level of 10-6
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. The 96-Well plates have an orange embossed alphanumeric code for quick and easy well identification.
The hydrophilic surface has freely accessible amino groups in addition to carboxyl and hydroxyl groups for a protein-like surface, allowing cells to directly attach and spread out. Suitable for reduced serum cultivation. All plates are made of virgin polystyrene. Sterilized by beta irradiation according to Ph. Eur. and USP 29, to a Sterility Assurace Level of 10-6.
Ordering information: All plates are individually wrapped with lid in packs of 50.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty