Would you like to log in again?
Lab Frame Foot, Chemglass
Supplier: Chemglass
CG-9274-01 CG-9274-S-01
80063-668EA 48.56 USD InStock
80063-668 89094-302
Lab Frame Foot, Chemglass
Support Rods and Frames
Lab frame feet in two different versions.
  • Permanent lattice mounting to bench tops, floors or walls
  • Accommodates 1/2” (13 mm) diameter rod
  • Available in stainless steel electro-polished finish
Supplied with three screws.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty