Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunc® Centrifuge Tubes, Polycarbonate, Thermo Scientific
Supplier: Thermo Scientific
Nunc®
347708
89094-330CS 754.78 USD InStock
89094-330
Nunc® Centrifuge Tubes, Polycarbonate, Thermo Scientific
Tubes Centrifuge Tubes
Sterile, conical-bottom centrifuge tubes can withstand centrifugal forces upto 6000 ×g.
  • Tubes are autoclavable
  • Total volume: 11 ml
Rim seal polypropylene caps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty