Would you like to log in again?
Pasteur and Serum Pipettes, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
612301 612362 700370 612361 700361
82051-088CS 300 USD InStock
82051-088 89094-392 89094-394 82051-090 82050-498
Pasteur and Serum Pipettes, Greiner Bio-One
Pipettes Pasteur/Transfer Pipettes
Greiner Bio-One Pasteur and Serum Pipettes are ideally suited for the rapid transfer of liquids.
  • Sterile or nonsterile
  • Provided with or without graduations
  • Available in a range of maximum fill volumes
Pipettes are available sterile or nonsterile.
Product numbers 700370 and 700361 are designed without graduations.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty