Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Bel-Art 4-Color Wash Bottles, Right-to-Know and Safety-Labeled, Wide Mouth, Bel-Art Products, a part of SP

Supplier: Bel-Art

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

F11716-0014 F11716-0020 F11716-0004 F11716-0010 F11716-0001 F11732-0019 F11716-0017 F11716-0050 F11716-0015 F11716-0003 F11716-0009 F11716-0008 F11732-0011 F11732-0009 F11732-0001 F11716-0011 F11732-0008 F11716-0007 F11732-0015 F11716-0019 F11716-0013 F11716-0012 F11716-0016 F11716-0002 F11732-0004
89141-022CS 226.67 USD InStock
89141-022 89141-024 89094-616 89094-618 89094-634 89094-876 89094-614 89094-630 89094-872 89094-632 89094-870 89141-036 89094-628 89094-606 89094-608 89094-602 89094-624 89094-868 89094-626 89094-604 89094-620 89094-862 89094-622 89094-864 89141-032
SP Bel-Art 4-Color Wash Bottles, Right-to-Know and Safety-Labeled, Wide Mouth, Bel-Art Products, a part of SP
Bottles Wash Bottles
Right-to-Know safety-labeled wash bottles feature a 53 mm (2") wide mouth for easy refilling.

  • Pre-printed with chemical name and formula
  • Leakproof
  • Translucent LDPE construction
  • Color-coded polypropylene closures for quick identification

Bottles are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment

*Note: Use LYOB only with solvents compatible with LDPE.

Leak-proof, these translucent* LDPE bottles have color-coded polypropylene closures to aid in quick and easy identification.

Certifications: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.

Ordering information: Bottles are packaged 4/bag. The Assortment item includes 6/bag, with one each of Acetone, Bleach, Distilled Water, Ethanol, Isopropanol, and Label Your Own Bottle (LYOB).

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty