Would you like to log in again?
VWR® Traceable® High-Accuracy Refrigerator/Freezer Digital Thermometer
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Traceable®
93677-81 VC2046 VC1046 93677-80 90011-90 93675-76 90008-65 VC4238
10171-458EA 155.18 USD InStock
10171-458 89094-750 10171-574 89094-748 10208-392 10171-294 89094-746 10205-164
VWR® Traceable® High-Accuracy Refrigerator/Freezer Digital Thermometer
Thermometers Digital Thermometers
Perfect for monitoring freezers, heating blocks, incubators, refrigerators, and water baths.
  • Traceable
  • Temperature in °C and °F
  • Chemical-resistant, high-impact ABS plastic casing
  • Continuous visual and audio alerts
  • Does not affect seal of closed doors
  • One-button self-calibration
Display case is made of high-impact, chemical-resistant ABS plastic
For thermometers 89094-746 and 89094-748, each sensor probe is sealed in a bottle filled with GRAS nontoxic glycol. The solution insulates the sensor from rapid temperature changes, such as when refrigerator door is opened. Bottle dimensions: 2.5x6.4 cm (1x21/2"). Thermometer 89094-750 has two chemical-resistant, waterproof sensor probes. Probe diameter: 5mm (3/16"). Probe length: 1.9 cm (3/4").
Display simultaneously shows maximum, minimum, and current probe temperatures, as well as room temperature. Alarm feature provides continuous visual and audio alerts when temperature rises above or falls below a programmable set point, even if temperature returns to non-alarm range. Dual probe thermometers provide individual readings and alerts for each probe. Sensor and 3m (10') micro-cable continue to perform accurately when submerged in water. Micro-cable permits refrigerator doors to close on it without affecting the seal. Thermometers feature one-button self-calibration.
Certifications: Thermometers fulfill all CDC thermometer and vaccine thermometer requirements. A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: All thermometers are supplied with two AA alkaline batteries, stand, wall mount, Velcro® mounting strips, and magnetic mounting strips.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty