Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Bel-Art Wash Bottles, 4-Color, Right-to-Know, Safety-Vented and Safety-Labeled, Wide Mouth, Bel-Art Products, a part of SP

Supplier: Bel-Art

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SCIENCEWARE®
F11816-0001 F11808-0004 F11832-0004 F11816-0008 F11808-0008 F11816-0011 F11832-0011 F11808-0015 F11832-0001 F11816-0019 F11816-0015 F11832-0019 F11808-0009 F11816-0009 F11816-0004 F11808-0001 F11832-0008 F11808-0019 F11808-0011 F11808-0050 F11816-0050 F11832-0009
89094-638CS 421.71 USD InStock
89094-638 89141-026 89094-902 89141-038 89094-904 89141-028 89094-646 89094-888 89094-900 89094-648 89094-894 89094-641 89094-896 89094-654 89094-886 89094-890 89094-880 89094-650 89094-892 89094-882 89141-040 89141-030
SP Bel-Art Wash Bottles, 4-Color, Right-to-Know, Safety-Vented and Safety-Labeled, Wide Mouth, Bel-Art Products, a part of SP
Bottles Wash Bottles
A unique valve prevents pressure build-up inside these Right-to-Know Safety-Labeled Wash Bottles, eliminating potentially dangerous drips.

  • Wide mouth for easy refilling
  • Leakproof
  • Translucent LDPE construction
  • Color-coded polypropylene closures for quick identification

This valve allows dispensing in an upright or inverted position.

Bottles feature a 53 mm (2") wide mouth for easy refilling. They are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Leak-proof, these translucent* LDPE bottles have color-coded polypropylene closures to aid in quick and easy identification.

*Note: Use LYOB only with solvents compatible with LDPE.

Certifications: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.

Ordering information: The 250 and 500 ml bottles are packaged four per bag. The 1000 ml bottles are packaged two per bag. The Assortment item includes six per bag, with one each of Acetone, Bleach, Distilled Water, Ethanol, Isopropanol, and Label Your Own Bottle (LYOB).

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty