Would you like to log in again?
Female Luer Lock Vacuum Adapters, Chemglass
Supplier: Chemglass
CG-1049-F-24 CG-1049-F-14
89094-918EA 240.36 USD InStock
89094-918 89094-920
Female Luer Lock Vacuum Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters have a 1/4"–28 thread that accepts a female luer lock adapter. This provides a convenient way to connect solid phase extraction (SPE) cartridges, tubes, or syringes with a male luer lock fitting to a standard male taper inner joint.
  • Hose connection is 10 mm O.D. at the largest serration
The adapter enables easier sample concentration and clean-up while reducing the chances of cross-contamination.
Delivery information: Supplied complete with one 1/4"–28 Female to Female luer lock adapter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty