Would you like to log in again?
VWR® TraceClean® Wide Mouth Packers, HDPE
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® TraceClean®
QLPW15900299 QLPW15900111 QLPW15900115 QLPW15900113 QLPW15900301 QLPW15900117 QLPW15900209 QLPW15900297 QLPW15900205
89095-040CS 526.65 USD InStock
89095-040 89095-034 89095-043 89095-037 89095-049 89095-046 89095-035 89095-047 89095-028
VWR® TraceClean® Wide Mouth Packers, HDPE
Bottles Plastic Bottles
Designed for the collection of water samples for testing metals and a variety of inorganic analytes such as cyanide, sulfide, sulfate, chloride, and ion chromatography analytes.
  • HDPE construction
  • Wide-mouthed design
  • Supplied with polyfoam-lined caps
Both the Processed (PC) and Quallity-Assured® (QA) containers follow EPA cleaning procedures according to the OSWER Directive No. 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA containers include a Certificate of Analysis in every case, as well as bar codes containing both the lot number and individual container number, ensuring complete traceability. Not Processed (NPC) containers do not undergo the EPA cleaning process. All container cases are custody sealed to ensure product integrity.
Ordering information: Constructed of sturdy HDPE, packers are supplied with polyfoam-lined closures.
Packaging: Packaged in bulk.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty