Would you like to log in again?
VWR® Erlenmeyer Flasks, Polycarbonate, Sterile
Supplier: VWR
VWR®
431550 431548 431553 431547 431544 FBC2000S-VWR 431551 431545 431543 431555 431552 431540 431554 431542 FPC3000S-VWR FBC3000S-VWR 431546 431541 FPC2000S-VWR 431549
89095-282CS 931.85 USD InStock
89095-282 89095-260 89095-272 10126-438 89095-280 89095-270 89095-264 89095-286 89095-276 89095-284 10126-408 89095-262 89095-274 89095-268 89095-258 89095-266 89095-288 89095-278 10126-412 10126-410
VWR® Erlenmeyer Flasks, Polycarbonate, Sterile
Flasks Erlenmeyer Flasks
These flasks are optimally designed for use in all aerobic/anaerobic suspension cultures.
  • Autoclavable once
  • Virgin, medical-grade resin
  • Chemically inert and non-leaching
  • Wide temperature tolerance
  • Uniform thickness
  • Molded-in graduations
These flasks can be used for storage, media preparation, mammalian, microbial, and plant cell culture and related applications.
Flasks are manufactured from virgin, medical-grade resin that retains its clarity after being exposed to gamma radiation. They are inert, non-leaching, and provide a wide temperature tolerance and uniform thickness. They are also tested with E. coli cultures for improved nucleic acid yield. Flasks are available with either a flat or baffled base, and they will fit all standard shaking incubator clamps. Molded-in graduations are included on all sizes.
The 125mL, 250mL, 500mL and 1L flasks are available with a vented cap with a 0.22µm pore-size PTFE membrane for aerobic (sterile air exchange) and a solid cap for anaerobic (no air transfer) applications. The 2L and 3L flasks come with a convenient vented duo cap that makes switching caps easy; simply remove the dustguard cover to allow sterile exchange, and then replace the cover when ready.
Certifications: Flasks and caps are autoclavable once and sterility tested to SAL 10-6. Certified pyrogen- and DNase/RNase-free.
Packaging: Flasks are individually wrapped.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty